DL-Serine ≥98%

Synonyms: (±)-Serine, H-DL-Ser-OH, DL-2-Amino-3-hydroxypropionic acid

S4375-100G S4375-1KG
220209-369EA 114.58 USD
220209-369 220209-370
DL-Serine ≥98%
DL-Serine
Formula: C₃H₇NO₃
MW: 105.09 g/mol
Boiling Pt: 395 °C (1013 hPa)
Melting Pt: 243 - 244 °C
Density: 1.543 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00064223
CAS Number: 302-84-1
EINECS: 206-130-6
Merck Index: 13,08534

Order Now

Specification Test Results

Assay 98% (TLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR