Sodium carbonate monohydrate ≥99.5%

Warning

S4132-1KG S4132-500G
220209-338EA 73.23 USD
220209-338 220209-339
Sodium carbonate monohydrate ≥99.5%
Sodium carbonate monohydrate
Formula: Na₂CO₃·1H₂O
Melting Pt: 107.85 °C
Density: 2.25 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149177
CAS Number: 5968-11-6
EINECS: 207-838-8

Order Now

Specification Test Results

Assay 99.5%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR