Sodium iodate

Danger

S4007-500G S4007-1KG S4007-100G
220208-719EA 154.38 USD
220208-719 220208-718 220208-717
Sodium iodate
Sodium iodate
Formula: INaO₃
MW: 197.89 g/mol
Melting Pt: 420…430 °C
Density: 3.56 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003533
CAS Number: 7681-55-2
EINECS: 231-672-5
UN: 1479
ADR: 5.1,II
Merck Index: 13,08704
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=806173453dcec1d2f791367d269d662d3459149a&vid=16d5efbf45a7cc1cd6399272211e2455e47a2e4b&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR