Squalene

S3626-1L S3626-100ML S3626-500ML S3626-10ML
220208-996EA 978.46 USD
220208-996 220208-994 220208-997 220208-995
Squalene
Squalene
Formula: C₃₀H₅₀
Boiling Pt: 429 °C (1013 hPa)
Melting Pt: –5 °C
Density: 0.86 g/cm³ (20 °C)
Flash Pt: 200 °C
MDL Number: MFCD00008912
CAS Number: 111-02-4
EINECS: 203-826-1
Merck Index: 13,08847

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR