Spermidine trihydrochloride ≥98%

S2501-1G S2501-25G S2501-5G
220208-083EA 55.68 USD
220208-083 220208-084 220208-085
Spermidine trihydrochloride ≥98%
Spermidine trihydrochloride
Formula: C₇H₂₂Cl₃N₃
MW: 254.63 g/mol
Melting Pt: 256 - 258 °C
Storage Temperature: Freezer
MDL Number: MFCD00012918
CAS Number: 334-50-9
EINECS: 206-379-0
Merck Index: 12,08893

Order Now

Specification Test Results

Assay 98% TLC

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR