di-Sodium succinate hexahydrate ≥99%, ReagentPlus®

S2378-100G S2378-500G S2378-1KG
220208-963EA 31.15 USD
220208-963 220208-965 220208-964
di-Sodium succinate hexahydrate ≥99%, ReagentPlus®
di-Sodium succinate hexahydrate
Formula: NaOOCCH₂CH₂COONa·6H₂O
MW: 270.14 g/mol
Melting Pt: 300 °C
Density: 1.529 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149117
CAS Number: 6106-21-4
EINECS: 205-778-7
Merck Index: 13,08754

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR