Sodium nitrite, ReagentPlus®

Danger

Synonyms: Diazotizing salts

S2252-500G S2252-2.5KG
220208-942EA 38.34 USD
220208-942 220208-941
Sodium nitrite, ReagentPlus®
Sodium nitrite
Formula: NaNO₂
MW: 69 g/mol
Boiling Pt: 320 °C (1013 hPa)
Melting Pt: 284 °C
Density: 2.168 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00011118
CAS Number: 7632-00-0
EINECS: 231-555-9
UN: 1500
ADR: 5.1,III
Merck Index: 13,08721

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR