Sodium azide ≥99.5%, ReagentPlus®

Danger

S2002-25G S2002-100G S2002-500G S2002-2KG
220208-909EA 48.65 USD
220208-909 220208-908 220208-615 220208-910
Sodium azide ≥99.5%, ReagentPlus®
Sodium azide
Formula: NaN₃
Melting Pt: 275 °C
Density: 1.85 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003536
CAS Number: 26628-22-8
EINECS: 247-852-1
UN: 1687
ADR: 6.1,II
Merck Index: 13,08653

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR