Sorbic acid

Warning

Synonyms: (E,E)-2,4-Hexadienoic acid, (2E,4E)-2,4-Hexadienoic acid, trans,trans-2,4-Hexadienoic acid, 2,4-Hexadienoic acid, (2E,4E)-Hexa-2,4-dienoic acid

S1626-500G S1626-100G S1626-250G
220208-765EA 77.54 USD
220208-765 220208-763 220208-764
Sorbic acid
Sorbic acid
Formula: CH₃CH=CHCH=CHCOOH
MW: 112.13 g/mol
Boiling Pt: 228 °C (1013 hPa)
Melting Pt: 132…135 °C
Density: 1.204 g/cm³ (20 °C)
Flash Pt: 127 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00002703
CAS Number: 110-44-1
EINECS: 203-768-7
Merck Index: 13,08793
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=440c50b4fc52d9ff0b52afa7ba596c860f95e0e7&vid=f9b0ff409010208781696e9ca3314c02aaf1af36&prid=web&clid=SAPDAM

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR