Sodium dihydrogen phosphate ≥99.0%, ReagentPlus®

Synonyms: Sodium dihydrogenphosphate (granular), Sodium dihydrogen orthophosphate, Monobasic Sodium Phosphate, Sodium phosphate monobasic

S0751-100G S0751-3KG S0751-1KG S0751-500G
220208-457EA 23.85 USD
220208-457 220208-459 220208-458 220208-460
Sodium dihydrogen phosphate ≥99.0%, ReagentPlus®
Sodium dihydrogen phosphate
Formula: NaH₂PO₄
MW: 119.98 g/mol
Melting Pt: ∼650 °C
Density: 0.9996 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003527
CAS Number: 7558-80-7
EINECS: 231-449-2
REACH: 01-2119489796-13
Merck Index: 12,08806

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR