Sodium sulfite

S0505-2.5KG S0505-250G
220207-586EA 105.85 USD
220207-586 220207-587
Sodium sulfite
Sodium sulphite
Formula: Na₂SO₃
MW: 126.04 g/mol
Melting Pt: > 500 °C
Density: 2.63 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003503
CAS Number: 7757-83-7
EINECS: 231-821-4
REACH: 01-2119537420-49
Merck Index: 13,08757

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR