Riboflavine (vitamin B2)

R4500-1KG R4500-5G R4500-100G R4500-25G R4500-500G
220207-774EA 1155 USD
220207-774 220207-777 220207-773 220207-775 220207-776
Riboflavine (vitamin B2)
Riboflavine (vitamin B2)
Formula: C₁₇H₂₀N₄O₆
Melting Pt: 280 °C
Storage Temperature: Ambient
MDL Number: MFCD00005022
CAS Number: 83-88-5
EINECS: 201-507-1
Merck Index: 12,08367

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR