L(+)-Ribose

Synonyms: (2S,3S,4S)-2,3,4,5-Tetrahydroxypentanal, L-(+)-Ribose

R4377-1G R4377-5G
220207-764EA 699.08 USD
220207-764 220207-765
L(+)-Ribose
L(+)-Ribose
Formula: C₅H₁₀O₅
MW: 150.13 g/mol
Boiling Pt: 331 °C (1013 hPa)
Melting Pt: 86 - 87 °C
Storage Temperature: Ambient
MDL Number: MFCD00167010
CAS Number: 24259-59-4
EINECS: 246-110-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR