1,4-Dihydroxy-9,10-anthraquinone

Synonyms: 1,4-Dihydroxyanthracene-9,10-dione, Quinizarin, 1,4-Dihydroxyanthraquinone

Q906-5G Q906-100G
220206-814EA 36.14 USD
220206-814 220206-813
1,4-Dihydroxy-9,10-anthraquinone
1,4-Dihydroxy-9,10-anthraquinone
Formula: C₁₄H₈O₄
MW: 240.22 g/mol
Boiling Pt: 465 °C (1013 hPa)
Melting Pt: 202 °C
Density: 1.278 g/cm³ (20 °C)
MDL Number: MFCD00001209
CAS Number: 81-64-1
EINECS: 201-368-7
Merck Index: 12,08250

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR