Quinine hemisulfate monohydrate (synthetic) ≥90% (by HPLC)

Synonyms: Quinine sulphate, Quinine sulphate (2:1) (salt) dihydrate

Q1250-50G Q1250-5G Q1250-10G
220206-912EA 232.89 USD
220206-912 220206-913 220206-911
Quinine hemisulfate monohydrate (synthetic) ≥90% (by HPLC)
Quinine hemisulfate monohydrate
Formula: C₂₀H₂₄N₂O₂·0.5H₂SO₄·H₂O
MW: 391.47 g/mol (373.46 anhy)
Melting Pt: 219 °C
Density: 1.36 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00150792
CAS Number: 6119-70-6
EINECS: 212-359-2
Merck Index: 12,03822

Order Now

Specification Test Results

Assay 90%
(HPLC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR