Tween® 20 (Polysorbate) for molecular biology

Synonyms: Polyoxyethylene sorbitan monolaurate

P9416-50ML P9416-100ML
220205-878EA 38.95 USD
220205-878 220205-877
Tween® 20 (Polysorbate) for molecular biology
Tween® 20 (Polysorbate)
Formula: C₅₈H₁₁₄O₂₆
MW: 1227.53 g/mol
Density: 1.1 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00165986
CAS Number: 9005-64-5
EINECS: 500-018-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR