Ethyl palmitate ≥99%

Synonyms: Hexadecanoic acid ethyl ester, Hexadecylic acid ethyl ester, Ethyl hexadecanoate, Palmitic acid ethyl ester

P9009-25G P9009-5G
220205-500EA 178.46 USD
220205-500 220205-501
Ethyl palmitate ≥99%
Ethyl palmitate
Formula: C₁₈H₃₆O₂
MW: 284.48 g/mol
Boiling Pt: 342 °C (1013 hPa)
Melting Pt: 24 °C
Density: 0.86 g/cm³ (20 °C)
MDL Number: MFCD00008996
CAS Number: 628-97-7
EINECS: 211-064-6

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR