3-Pentanol ≥98%

P8025-500ML-A P8025-25ML-A P8025-100ML-A
220205-256EA 215.38 USD
220205-256 220205-255 220205-254
3-Pentanol ≥98%
3-Pentanol
Formula: C₅H₁₂O
MW: 88.15 g/mol
Boiling Pt: 116 °C (1013 hPa)
Melting Pt: -8 °C
Density: 0.82 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00004574
CAS Number: 584-02-1
EINECS: 209-526-7
UN: 1105
ADR: 3,III
Merck Index: 13,07197
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=2ec7192926b50615dfac930445c63174c6221a44&vid=a2f1fa3f1ffa1ecbfb6a5dd1d0f29da27d1d8737&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR