Tetrasodium diphosphate

Warning

Synonyms: Sodium pyrophosphate tetrabasic, Tetrasodium diphosphate, Sodium diphosphate, Sodium pyrophosphate, tetra-Sodium pyrophosphate

P8010-500G P8010-2.5KG
220205-244EA 75.08 USD
220205-244 220205-243
Tetrasodium diphosphate
Tetrasodium diphosphate
Formula: Na₄O₇P₂
MW: 265.9 g/mol
Melting Pt: 988 °C
Density: 2.534 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00003513
CAS Number: 7722-88-5
EINECS: 231-767-1
Merck Index: 13,09312

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR