1-Phenyl-2-thiourea ≥98%

Danger

Synonyms: N-Phenylthiourea, 1-Phenylthiourea

P7629-10G P7629-25G P7629-100G
220205-022EA 58.48 USD
220205-022 220205-023 220205-021
1-Phenyl-2-thiourea ≥98%
1-Phenyl-2-thiourea
Formula: C₇H₈N₂S
MW: 152.22 g/mol
Boiling Pt: 295…297 °C (1013 hPa)
Melting Pt: 84…85 °C
Density: 1.33 g/cm³ (20 °C)
MDL Number: MFCD00004933
CAS Number: 103-85-5
EINECS: 203-151-2
UN: 2811
ADR: 6.1,I
Merck Index: 12,07468

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR