Toluene-α-sulfonyl fluoride ≥98.5% (by GC)

Danger

Synonyms: PMSF, Phenylmethanesulphonyl fluoride, Phenylmethylsulphonyl fluoride, α-Toluenesulphonyl fluoride

P7626-1G P7626-250MG P7626-25G P7626-100G P7626-5G
220205-014EA 78.92 USD
220205-014 220205-015 220205-016 220205-013 220205-017
Toluene-α-sulfonyl fluoride ≥98.5% (by GC)
Toluene-α-sulphonyl fluoride
Formula: C₇H₇FO₂S
MW: 174.2 g/mol
Boiling Pt: 80…85 °C (0.3 mmHg)
Melting Pt: 91…92 °C
Storage Temperature: Ambient
MDL Number: MFCD00007424
CAS Number: 329-98-6
EINECS: 206-350-2
UN: 2923
ADR: 8,III

Order Now

Specification Test Results

Assay 98.5% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR