2-Pyrrolecarbaldehyde

Synonyms: 1H-Pyrrole-2-carboxaldehyde, Pyrrole-2-carboxaldehyde, 1H-Pyrrole-2-carbaldehyde, Pyrrole-2-carbaldehyde

P73404-5G P73404-100G P73404-25G
220203-488EA 81.63 USD
220203-488 220203-486 220203-487
2-Pyrrolecarbaldehyde
2-Pyrrolecarbaldehyde
Formula: C₅H₅NO
MW: 95.1 g/mol
Boiling Pt: 217 °C (1013 hPa)
Melting Pt: 43…46 °C
MDL Number: MFCD00005217
CAS Number: 1003-29-8
EINECS: 213-705-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR