Pyrazinamide

P7136-100G P7136-25G P7136-10G
220204-863EA 350.77 USD
220204-863 220204-865 220204-864
Pyrazinamide
Pyrazinamide
Formula: C₅H₅N₃O
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 188 - 191 °C
MDL Number: MFCD00006132
CAS Number: 98-96-4
EINECS: 202-717-6
Merck Index: 12,08140

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR