3-Pyridylmethanol

Warning

Synonyms: 3-(Hydroxymethyl)pyridine, 3-Pyridinemethanol, 3-Pyridylcarbinol, Pyridine-3-methanol, 3-Pyridylmethanol

P66807-5G P66807-100G
220204-718EA 37.72 USD
220204-718 220204-717
3-Pyridylmethanol
3-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 266 °C (1013 hPa)
Melting Pt: -7 °C
Density: 1.1357 g/cm³ (20 °C)
MDL Number: MFCD00006407
CAS Number: 100-55-0
EINECS: 202-864-6
Merck Index: 12,06614

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR