2-Pyridylmethanol

Warning

Synonyms: 2-Pyridylmethanol, 2-Pyridylcarbinol, 2-Pyridinemethanol, Pyridin-2-ylmethanol, 2-(Hydroxymethyl)pyridine, Pyridine-2-methanol

P66602-100G P66602-25G
220204-704EA 71.4 USD
220204-704 220204-705
2-Pyridylmethanol
2-Pyridylmethanol
Formula: C₆H₇NO
MW: 109.13 g/mol
Boiling Pt: 223 °C (1013 hPa)
Melting Pt: –6 °C
Density: 1.1317 g/cm³ (20 °C)
MDL Number: MFCD00006348
CAS Number: 586-98-1
EINECS: 209-592-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR