Pyridine-2,3-dicarboxylic anhydride

Warning

P64405-100G P64405-5G P64405-25G
220204-615EA 175.54 USD
220204-615 220204-617 220204-616
Pyridine-2,3-dicarboxylic anhydride
Pyridine-2,3-dicarboxylic anhydride
Formula: C₇H₃NO₃
Boiling Pt: 360 °C (1013 hPa)
Melting Pt: 146 °C
MDL Number: MFCD00005915
CAS Number: 699-98-9
EINECS: 211-834-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR