Pyridine-2,4-dicarboxylic acid monohydrate ≥98%, Sigma-Aldrich®

Warning

Synonyms: Lutidinic acid monohydrate, 2,4-Pyridinedicarboxylic acid monohydrate

P63395-5G P63395-1G
220204-576EA 83.78 USD
220204-576 220204-575
Pyridine-2,4-dicarboxylic acid monohydrate ≥98%, Sigma-Aldrich®
Pyridine-2,4-dicarboxylic acid monohydrate
Formula: C₇H₅NO₄·1H₂O
MW: 185.14 g/mol
CAS Number: 207671-42-9
EINECS: 207-892-2

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR