Pyridine-2,3-dicarboxylic acid

Synonyms: Quinolinic acid

P63204-25G P63204-100G
220204-574EA 32.52 USD
220204-574 220204-573
Pyridine-2,3-dicarboxylic acid
Pyridine-2,3-dicarboxylic acid
Formula: C₇H₅NO₄
MW: 167.12 g/mol
Boiling Pt: 357 °C (1013 hPa)
Melting Pt: 188 - 190 °C
Storage Temperature: Ambient
MDL Number: MFCD00006295
CAS Number: 89-00-9
EINECS: 201-874-8
Merck Index: 12,08258

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR