Tween® 80 (Polysorbate)

P6224-1GA P6224-500ML
220204-549EA 323.32 USD
220204-549 220204-550
Tween® 80 (Polysorbate)
Tween® 80 (Polysorbate)
Melting Pt: 100 °C
Flash Pt: 113 °C (closed cup)
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR