Prednisolone

Synonyms: Prednisolonum

P6004-25G P6004-100MG P6004-1G P6004-5G P6004-10G
220204-514EA 693.32 USD
220204-514 220204-511 220204-513 220204-515 220204-512
Prednisolone
Prednisolone
Formula: C₂₁H₂₈O₅
MW: 360.45 g/mol
Boiling Pt: 571 °C (1013 hPa)
Melting Pt: 246 °C
Density: 1.212 g/cm³ (20 °C)
MDL Number: MFCD00003649
CAS Number: 50-24-8
EINECS: 200-021-7

Order Now

Specification Test Results

Assay 99%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR