p-Phenylenediamine

Danger

Synonyms: 1,4-Diaminobenzene, PPD, 1,4-Phenylenediamine

P6001-250G P6001-1KG P6001-100G P6001-50G
220204-509EA 119.54 USD
220204-509 220204-508 220204-507 220204-510
p-Phenylenediamine
p-Phenylenediamine
Formula: C₆H₈N₂
MW: 108.14 g/mol
Boiling Pt: 267 °C (1013 hPa)
Melting Pt: 139…141 °C
Density: 1.251 g/cm³ (20 °C)
Flash Pt: 156 °C
Storage Temperature: Ambient
MDL Number: MFCD00007901
CAS Number: 106-50-3
EINECS: 203-404-7
UN: 1673
ADR: 6.1,III
Merck Index: 12,07439

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR