Pyrazine-2-carboxylic acid

Synonyms: Pyrazinecarboxylic acid, 2-Pyrazinecarboxylic Acid

P56100-100G P56100-25G
220204-223EA 98.43 USD
220204-223 220204-224
Pyrazine-2-carboxylic acid
Pyrazine-2-carboxylic acid
Formula: C₅H₄N₂O₂
MW: 124.1 g/mol
Boiling Pt: 313 °C (1013 hPa)
MDL Number: MFCD00006130
CAS Number: 98-97-5
EINECS: 202-718-1
Merck Index: 12,08143

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR