Pyrazine

P56003-10G P56003-5G P56003-25G
220204-215EA 46.2 USD
220204-215 220204-217 220204-216
Pyrazine
Pyrazine
Formula: C₄H₄N₂
MW: 80.09 g/mol
Boiling Pt: 115 °C (1013 hPa)
Melting Pt: 52 °C
Density: 1.0311 g/cm³ (60.9 °C)
MDL Number: MFCD00006122
CAS Number: 290-37-9
EINECS: 206-027-6
UN: 1325
ADR: 4.1,III
Merck Index: 13,08047

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR