Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, ReagentPlus®

Synonyms: Dipotassium hydrogen phosphate trihydrate, Potassium hydrogen phosphate trihydrate, di-Potassium hydrogen phosphate trihydrate, Potassium phosphate dibasic trihydrate, di-Potassium hydrogen orthophosphate trihydrate

P5504-1KG P5504-500G P5504-100G P5504-5KG P5504-10KG
220204-165EA 212.26 USD
220204-165 220204-166 220204-163 220204-167 220204-164
Dipotassium hydrogen orthophosphate trihydrate ≥99.0%, ReagentPlus®
Dipotassium hydrogen orthophosphate trihydrate
Formula: K₂HPO₄·3H₂O
MW: 228.22 g/mol
Boiling Pt: ∼340 °C (1013 hPa)
Density: 2.504 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00149926
CAS Number: 16788-57-1
EINECS: 231-834-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR