Polyethylene glycol 1,450

Synonyms: PEG 1,450, Ethylene glycol polymer 1450, PEG 1450

P5402-1KG P5402-500G P5402-2.5KG
220204-126EA 114.15 USD
220204-126 220204-128 220204-127
Polyethylene glycol 1,450
Polyethylene glycol 1,450
Formula: H(OCH₂CH₂)nOH
MW: 1450 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00081839
CAS Number: 25322-68-3
EINECS: 500-038-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR