Propyl-4-hydroxybenzoate ≥99%

Warning

Synonyms: Propyl 4-hydroxybenzoate, Propyl parahydroxybenzoate, Propylparaben, 4-Hydroxybenzoic acid propyl ester

P53357-500G P53357-5G P53357-250G
220204-079EA 68.31 USD
220204-079 220204-080 220204-078
Propyl-4-hydroxybenzoate ≥99%
Propyl-4-hydroxybenzoate
Formula: C₁₀H₁₂O₃
MW: 180.2 g/mol
Boiling Pt: 292 °C (1013 hPa)
Melting Pt: 95 °C
Density: 1.28 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00002354
CAS Number: 94-13-3
EINECS: 202-307-7
Merck Index: 13,07958

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR