Propionyl chloride ≥98%

Danger

P51559-25G P51559-500G
220203-601EA 41.35 USD
220203-601 220203-602
Propionyl chloride ≥98%
Propionyl chloride
Formula: C₃H₅ClO
MW: 92.52 g/mol
Boiling Pt: 80 °C (1013 hPa)
Melting Pt: -94 °C
Density: 1.06 g/cm³ (20 °C)
Flash Pt: 6 °C
Storage Temperature: Ambient
MDL Number: MFCD00000745
CAS Number: 79-03-8
EINECS: 201-170-0
UN: 1815
ADR: 3,II
Merck Index: 12,08013

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR