Propiolic acid ≥95%

Danger

P51400-25G P51400-100G P51400-5G
220200-200EA 134.62 USD
220200-200 220200-199 220200-201
Propiolic acid ≥95%
Propiolic acid
Formula: C₃H₂O₂
Boiling Pt: 140 °C (1013 hPa)
Melting Pt: 18 °C
Density: 1.136 g/cm³ (20 °C)
Flash Pt: 58 °C
MDL Number: MFCD00004360
CAS Number: 471-25-0
EINECS: 207-437-8
UN: 2920
ADR: 8,II
Merck Index: 12,08006

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR