Propargyl amine ≥98%

Synonyms: Monopropagyl amine, Prop-2-ynylamine, 3-Aminopropyne, Prop-2-yn-1-amine

P50900-1G P50900-5G P50900-25G
220204-024EA 37.05 USD
220204-024 220204-026 220204-025
Propargyl amine ≥98%
Propargyl amine
Formula: C₃H₅N
MW: 55.08 g/mol
Boiling Pt: 83 °C (1013 hPa)
Density: 0.867 g/cm³ (16 °C)
MDL Number: MFCD00008198
CAS Number: 2450-71-7
EINECS: 219-513-8
UN: 3286
ADR: 3,II

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR