Propargyl alcohol ≥99%

Danger

Synonyms: 2-Propyn-1-ol, Prop-2-yn-1-ol

P50803-100ML P50803-1L P50803-500ML
220204-016EA 27.46 USD
220204-016 220204-017 220204-018
Propargyl alcohol ≥99%
Propargyl alcohol
Formula: C₃H₄O
MW: 56.06 g/mol
Boiling Pt: 114 °C (1013 hPa)
Melting Pt: –53 °C
Density: 0.9485 g/cm³ (20 °C)
Flash Pt: 33 °C
Storage Temperature: Ambient
MDL Number: MFCD00002912
CAS Number: 107-19-7
EINECS: 203-471-2
UN: 2929
ADR: 6.1,II
Merck Index: 12,07993

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR