1,3-Propanesultone

Danger

Synonyms: 1,2-Oxathiolane 2,2-dioxide

P50706-100G P50706-25G P50706-1KG
220204-008EA 183.08 USD
220204-008 220204-010 220204-009
1,3-Propanesultone
1,3-Propanesultone
Formula: C₃H₆O₃S
MW: 122.14 g/mol
Boiling Pt: 298 °C (1013 hPa)
Melting Pt: 31 °C
Density: 1.3996 g/cm³ (35 °C)
MDL Number: MFCD00005355
CAS Number: 1120-71-4
EINECS: 214-317-9
UN: 2811
ADR: 6.1,III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR