Piperonylic acid

Synonyms: 3,4-(Methylenedioxy)benzoic acid, Benzo[d][1,3]dioxole-5-carboxylic acid, 1,3-Benzodioxole-5-carboxylic acid

P49805-25G P49805-5G
220203-979EA 101.45 USD
220203-979 220203-980
Piperonylic acid
Piperonylic acid
Formula: C₈H₆O₄
MW: 166.13 g/mol
Boiling Pt: 180 °C (1 -rr)
Melting Pt: 232 - 235 °C
MDL Number: MFCD00005830
CAS Number: 94-53-1
EINECS: 202-342-8
Merck Index: 12,07630

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR