Piperonyl alcohol

Synonyms: Benzo[d][1,3]dioxol-5-ylmethanol, 3,4-(Methylenedioxy)benzyl alcohol, 1,3-Benzodioxol-5-ylmethanol

P49406-25G P49406-100G
220203-973EA 49.54 USD
220203-973 220203-972
Piperonyl alcohol
Piperonyl alcohol
Formula: C₈H₈O₃
MW: 152.15 g/mol
Boiling Pt: 282 °C (1013 hPa)
Melting Pt: 51 - 54 °C
MDL Number: MFCD00005836
CAS Number: 495-76-1
EINECS: 207-808-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR