Tween® 80 (Polysorbate)

P4780-100ML P4780-1GA P4780-500ML
220203-468EA 50.25 USD
220203-468 220203-469 220203-470
Tween® 80 (Polysorbate)
Tween® 80 (Polysorbate)
Boiling Pt: ∼100 °C (1013 hPa)
Melting Pt: –21 °C
Density: 1.062 g/cm³ (20 °C)
Flash Pt: >149 °C
Storage Temperature: Ambient
MDL Number: MFCD00082107
CAS Number: 9005-65-6
EINECS: 500-019-9
Merck Index: 13,07664

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR