Pentaerythritol ≥98%

P4755-1KG-A P4755-500G-A
220203-897EA 69.68 USD
220203-897 220203-898
Pentaerythritol ≥98%
Pentaerythritol
Formula: C₅H₁₂O₄
MW: 136.15 g/mol
Boiling Pt: 380 °C (1013 hPa)
Melting Pt: 260 - 264 °C
Density: 1.511 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00004692
CAS Number: 115-77-5
EINECS: 204-104-9
Merck Index: 12,07245

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR