Piperazine, ReagentPlus®

Danger

P45907-100G P45907-1KG P45907-5G P45907-500G
220203-838EA 26.62 USD
220203-838 220203-839 220203-841 220203-840
Piperazine, ReagentPlus®
Piperazine
Formula: C₄H₁₀N₂
Boiling Pt: 146 °C (1013 hPa)
Melting Pt: 44…46 °C
Density: 1.1 g/cm³ (20 °C)
Flash Pt: 65 °C
Storage Temperature: Ambient
MDL Number: MFCD00005953
CAS Number: 110-85-0
EINECS: 203-808-3
UN: 2579
ADR: 8,III
Merck Index: 13,07545

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR