3,3-Dimethylbutanone ≥98%

Danger

Synonyms: 1,1,1-Trimethylacetone, Methyl tert-butyl ketone, Pinacolone, tert-Butyl methyl ketone, 3,3-Dimethyl-2-butanone

P45605-500ML P45605-100ML
220203-828EA 316.62 USD
220203-828 220203-827
3,3-Dimethylbutanone ≥98%
3,3-Dimethylbutanone
Formula: C₆H₁₂O
MW: 100.16 g/mol
Boiling Pt: 106 °C (1013 hPa)
Melting Pt: -53 °C
Density: 0.803 g/cm³ (20 °C)
Flash Pt: 5 °C
Storage Temperature: Ambient
MDL Number: MFCD00008846
CAS Number: 75-97-8
EINECS: 200-920-4
UN: 1993
ADR: 3,II
Merck Index: 12,07594

Order Now

Specification Test Results

Assay 98%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR