cis-Platin

Synonyms: Platinol, cis-Diaminodichloroplatinum(II), cis-Diamminedichloroplatinum (II), cis-Dichlorodiammine platinum(II), Cisplatin, cis-Diamineplatinum(II) dichloride

P4394-100MG P4394-1G P4394-250MG P4394-25MG
220203-750EA 123.68 USD
220203-750 220203-751 220203-752 220203-753
cis-Platin
cis-Platin
Formula: H₆Cl₂N₂Pt
MW: 300.05 g/mol
Density: 3.738 g/cm³ (20 °C)
MDL Number: MFCD00011623
CAS Number: 15663-27-1
EINECS: 239-733-8
UN: 3288
ADR: 6.1,II
Merck Index: 13,02341

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR