Phloroglucinol dihydrate ≥97%

Warning

Synonyms: 1,3,5-Trihydroxybenzene dihydrate, Phloroglucinol, Hydrate

P38005-25G P38005-100G
220203-434EA 54.6 USD
220203-434 220203-433
Phloroglucinol dihydrate ≥97%
Phloroglucinol dihydrate
Formula: C₆H₆O₃·2H₂O
MW: 162.14 g/mol
Melting Pt: 218…221 °C
Storage Temperature: Ambient
MDL Number: MFCD00149090
CAS Number: 6099-90-7
EINECS: 203-611-2
Merck Index: 13,07413

Order Now

Specification Test Results

Assay 97%

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR