PIPES disodium salt

Synonyms: Piperazine-N,N'-bis-2-ethanesulphonic acid disodium salt, Piperazine-1,4-bis(2-ethanesulphonic acid) disodium salt, 2,2'-(Piperazine-1,4-diyl)bis(ethanesulphonic) acid disodium salt, PIPES-Na2, 1,4-Piperazinediethanesulphonic acid disodium salt, Disodium piperazine-1,4-diethanesulphonate

P3768-25G P3768-500G P3768-1KG P3768-100G P3768-250G
220203-416EA 50.02 USD
220203-416 220203-417 220203-414 220203-413 220203-415
PIPES disodium salt
PIPES disodium salt
Formula: C₈H₁₆N₂Na₂O₆S₂
MW: 346.34 g/mol
MDL Number: MFCD00064349
CAS Number: 76836-02-7
EINECS: 278-562-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR